Learn More
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
Brand:  Thermo Scientific™ 97002121
 View more versions of this product
Product Code. 10393175
  • 508.00€ / Pack of 900
Description
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 x 45 pieces
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2407310 Certificate TypeCertificate of Compliance Date19/06/2024 Catalog Number97002121
Lot Number2334910 Certificate TypeCertificate of Compliance Date10/01/2024 Catalog Number97002121
Lot Number2310810 Certificate TypeCertificate of Compliance Date08/05/2023 Catalog Number97002121
Lot Number2233410 Certificate TypeCertificate of Compliance Date13/12/2022 Catalog Number97002121
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Consumables for KingFisher™ mL Systems > KingFisher mL tube, (20x45 pcs)