Learn More
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
Brand:  Thermo Scientific™ 97002111
 View more versions of this product
Product Code. 10715156
  • 1008.00€ / Pack of 800
Description
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
800 pieces
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2130810 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number97002111
Lot Number2408410 Certificate TypeCertificate of Compliance Date21/11/2024 Catalog Number97002111
Lot Number2424110 Certificate TypeCertificate of Compliance Date30/10/2024 Catalog Number97002111
Lot Number2413010 Certificate TypeCertificate of Compliance Date28/08/2024 Catalog Number97002111
Lot Number2333310 Certificate TypeCertificate of Compliance Date10/01/2024 Catalog Number97002111
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Consumables for KingFisher™ mL Systems > KingFisher mL tip comb, 800 pcs