Learn More
Contec™ Bucket Wringers and Frames
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Brand: Contec™ 2756
778.54 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.- Choose from downpress, flat or curved styles
- Available in polypropylene or stainless steel
- Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
- Select wringer that complements mop
Specifications
Stainless Steel | |
Reusable | |
Contec™ VertiKlean MAX™ or QuickTask™ Flat Mops, 5 gal. (20 L) or 6.5 gal. (25 L) Bucket | |
Non-sterile |
Autoclavable | |
Ideal for cleanrooms and controlled environments | |
Stainless steel Flat Mop Wringer |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
29Hours
23Minutes
49Seconds
18New Offer Save Up to 92%