missing translation for 'onlineSavingsMsg'
Learn More

Proteintech CoraLite™488 TUNEL Assay Apoptosis Detection Kit (green)

A distinctive feature of apoptosis is degradation of DNA.

Brand:  Proteintech PF00006-50T

 View more versions of this product

Product Code. 17541176

  • 319.00€ / 50 tests
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The DNA fragments tend to be vary in length in integer multiples of 180 bp-200 bp, which is shown as a ladder-like pattern in agarose gel electrophoresis.
  • This kit uses the TUNEL method, using Terminal Deoxynucleotidyl Transferase (TdT) to catalyze the incorporation of CL488-dUTP at the 3'-OH end of the broken DNA of apoptotic cells.
  • CL488-dUTP-labeled DNA can be directly observed with a fluorescence microscope or quantified with a flow cytometer.
  • The TUNEL method can selectively detect apoptotic cells, but not necrotic cells or cells with DNA strand breaks caused by irradiation and drug treatment.
Specifications

Specifications

Emission: 515 nm
50 Tests
Flow Cytometry
Store at -20°C. Avoid exposure to light. Stable for 2 years after shipment. Avoid freeze- thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.