missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech CoraLite™594 TUNEL Assay Apoptosis Detection Kit
A remarkable feature of apoptosis is the degradation of chromosome DNA, which is a common phenomenon.
Brand: Proteintech PF00009-20T
This item is not returnable.
View return policy
Description
- The degradation is very specific and regular. DNA fragments break into different lengths are of 180 bp-200 bp, showing a specific ladder like ladder pattern in agarose gel electrophoresis.
- The kit uses TUNEL method and uses Terminal Deoxynucleotidyl Transferase(TdT) catalyzes the incorporation of CoraLite™594-dUTP into the 3'-OH terminal of the broken DNA of apoptotic cells.
- DNA labeled with CoraLite™594- dUTP can be observed directly by fluorescence microscope or quantified by flow cytometry.
- TUNEL can selectively detect apoptotic cells rather than necrotic cells or cells with DNA strand breaks caused by irradiation and drug therapy.
Specifications
Emission: 614 nm | |
20 Tests | |
Flow Cytometry | |
Store at -20°C. Avoid exposure to light. Stable for 2 years after shipment. Avoid freeze- thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction