Learn More
Corning™ 96-Well, CellBIND-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand: Corning™ 3340
This item is not returnable.
View return policy
Description
Specifications
Standard | |
0.32 cm2 | |
Black, Clear Bottom | |
96 | |
Sterile | |
360 μL | |
CellBIND-treated surface helps cells adapt to reduced serum or serum-free media and improves cell attachment for increased cell growth and yield. Requires no refrigeration or special handling, unlike standard coatings used to improve cell attachment. | |
5/Pk. |
With Lid | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
CellBIND | |
Flat Bottom (F-well) | |
50/Cs. | |
50/Cs. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
COR001 Lifescience Guid
File size: 7 MB
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Product Content Correction
Product Title
Corning™ 96-Well, CellBIND-Treated, Flat-Bottom Microplate > Black and Clear; Standard, with Lid; Yes; Corning CellBIND; Flat