missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cortisone Competitive ELISA Kit

Competitive ELISA

Specifications

Assay Sensitivity 29 pg/mL
Description Cortisone Competitive ELISA Kit
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA Kit
View More Specs

Description

Description

The Cortisone ELISA quantitates Cortisone in serum,saliva,urine,plasm,or dried fecal extracts.

Principle of the method

The Cortisone Competitive ELISA research-use-only kit is designed to quantitatively measure cortisone in extracted dried fecal samples,urine,saliva,plasma,and serum samples. This kit measures total cortisone in serum and plasma and in extracted fecal samples. A cortisone standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. A cortisone-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to cortisone to each well. After a two-hour incubation the plate is washed and TMB substrate is added. The substrate reacts with the bound cortisone-peroxidase conjugate to produce a colored product. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Cortisone (Kendall’s Compound E) is a glucocorticoid which is the product of cortisol (Compound F) deactivation by 11-beta-HSD2 (hydroxysteroid dehydrogenase), prohibiting receptor activation. The balance between cortisone and cortisol glucocorticoids helps to regulate the anti-inflammatory response. Cortisone is identical across all species.
Specifications

Specifications

29 pg/mL
HRP
ELISA Kit
Human
Colorimetric Microplate Reader
8.6%
RUO
-20°C
1 hr. 20 min.
Cortisone Competitive ELISA Kit
Fluorescence
Fecal Extract, Plasma, Saliva, Serum, Urine
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
11.9%
HRP
Inflammation
Chemical
2 hr. 30 min.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.