Learn More
Corning™ Costar™ Netwell™ Inserts
GREENER_CHOICE
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers.
Brand:  Corning™ 3477
473.91 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 10426651
  • 498.85€ / Pack of 48
Estimated Shipment: 20-05-2025
Description
Netwell Inserts have polyester (PET) mesh bottoms attached to a polystyrene ring or housing. They are used as tissue carriers, supports and strainers for culture of small organs, tissue slices, or explants at the air-media interface. They can be used to coarse filter tissue homogenates, cell suspensions, or microcarriers. Accessories allow them to be used as a handy carrier for immunocytochemical staining of tissue culture slices.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
Polystyrene
Documents
Product Information
Certificates
View accessory products
Product Title
Corning™ Costar™ Netwell™ Inserts > 15 mm; 74 μm