missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Antibody (SPM615) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-48116
This item is not returnable.
View return policy
Description
CPS1 Monoclonal specifically detects CPS1 in Human, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
CPS1 | |
Monoclonal | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CPS1 | |
Recombinant human CPS1 protein (Uniprot: P31327) | |
7 mL | |
Primary | |
This MAb recognizes a protein of 165kDa, identified as carbamoyl phosphate synthetase 1 (CPS1). This mitochondrial enzyme catalyzes synthesis of carbamoyl phosphate from ammonia and bicarbonate. This reaction is the first committed step of the urea cycle, which is important in the removal of excess urea from cells.Deficiency of CPS1 is an autosomal recessive disorder that causes hyperammonemia. CPS1 is a hepatocyte specific protein that localizes to the mitochondria of hepatocytes. It is a sensitive marker for distinguishing hepatocellular carcinomas (HCC) from other metastatic carcinomas as well as cholangio-carcinomas. HCC s occur primarily in the stomach, but they are also found in many other organs. CPS1 may also be a useful marker for intestinal metaplasia. Reportedly, strong expression of CPS1 correlates with smaller tumor size and longer patient survival. Occasionally, CPS1 is also found in gastric carcinomas as well as in a few other non-hepatic tumors. | |
Store at 4C. | |
IgG1 Îş |
Immunohistochemistry (Paraffin) | |
SPM615 | |
Immunohistochemistry-Paraffin | |
carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16 | |
Mouse | |
Protein A or G purified | |
RUO | |
1373 | |
Human, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CPS1 Antibody (SPM615) - IHC-Prediluted, Novus Biologicals™ > 7mL; Unlabeled
Spot an opportunity for improvement?Share a Content Correction