Learn More
STEMCELL Technologies CryoStor™ CS5
Animal-component-free, defined cryopreservation medium with 5% DMSO
Brand:  STEMCELL Technologies 07949
Encompass_Preferred
 View more versions of this product
Product Code. 17119931
  • 535.95€ / Pack of 5
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CryoStor™ CS5 is a uniquely formulated serum-free, animal component-free and defined cryopreservation medium containing 5% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS5 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS5 is cGMP-manufactured with USP grade components
Specifications
Specifications
CHO Cells, Mesenchymal Stem and Progenitor Cells, Other, Pluripotent Stem Cells
Cryopreservation
5 x 10 mL