Learn More
STEMCELL Technologies CryoStor™ CSB
Animal component-free and protein-free cryopreservation formulation
Brand:  STEMCELL Technologies 100-0238
Encompass_Preferred
 View more versions of this product
Product Code. 17147501
  • 4509.00€ / 500mL
Estimated Shipment: 27-01-2025
Description
  • CryoStor™ CSB is an intracellular-like base formulation from the CryoStor™ preservation media series
  • It is serum-free, animal component-free, and does not contain dimethyl sulfoxide (DMSO)
  • It is designed to improve post-thaw cell viability and function, and to reduce the level of cryopreservation-induced delayed-onset cell death
Specifications
Specifications
B Cells, CHO Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells
Cryopreservation
500 mL
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.