Learn More
Thermo Scientific™ Cryoware Permanent Markers and Pens
Identify samples and storage boxes permanently with these cryoware permanent markers and pens.
Brand: Thermo Scientific™ 6313-0010
25.83 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.
Features:
- Nunc™ and Nalgene™ cryoware markers come in a variety of colors for permanent handwritten identification of samples
- Use these markers by hand, or for higher throughput use them in the ID Scribe
Specifications
Extra Fine (Green is Fine) | |
Nalgene Cryoware Marker Set | |
4 |
Assorted (Red, Blue, Green, Black) | |
24 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7428894010 | Certificate of Compliance | 01/04/2025 | 6313-0010 |
7437557010 | Certificate of Compliance | 01/04/2025 | 6313-0010 |
7442290010 | Certificate of Compliance | 01/04/2025 | 6313-0010 |
7443066010 | Certificate of Compliance | 01/04/2025 | 6313-0010 |
7440044010 | Certificate of Compliance | 01/04/2025 | 6313-0010 |