missing translation for 'onlineSavingsMsg'
Learn More

CTP synthase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen CTP synthase
Concentration 0.16 mg/mL
Applications Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16855314
View Documents
Proteintech
15914-1-AP-20UL
20 ÎĽL
145.00€
20µL
Please to purchase this item. Need a web account? Register with us today!
16845314
View Documents
Proteintech
15914-1-AP-150UL
150 ÎĽL
410.00€
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

There are two genes encoding members of a new family of type II integral membrane proteins. Both are ubiquitously expressed, and tissue-specific alternative mRNA initiation and splicing generate at least two major isoforms of each protein, with the smaller isoforms being truncated at the N-terminus. These proteins are called nesprin-l and -2 for nuclear _envelope _spectrin repeat, as they are characterized by the presence of multiple, clustered spectrin repeats, bipartite nuclear localization sequences and a conserved C-terminal, single transmembrane domain. Transient transfection of EGFP-fusion expression constructs demonstrated their localization to the nuclear membrane with a novel C-terminal, TM-domain-containing sequence essential for perinuclear localization. Nesprin-l is developmentally regulated in both smooth and skeletal muscle and is relocalized from the nuclear envelope to the nucleus and cytoplasm during C2Cl2 myoblast differentiation. Nesprins may function as ′dystrophins of the nucleus′ to maintain nuclear organization and structura integrity
Specifications

Specifications

CTP synthase
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
Unconjugated
Rabbit
Mouse, Human, Rat
P17812, P70698
1503, 313560, 51797
CTP synthase Fusion Protein Ag8707
Primary
-20°C
CTPS1
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CTP synthase, CTP synthase 1, CTP synthetase 1, CTPS, CTPS 1, CTPS1, UTP ammonia ligase 1
Ctps, Ctps1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CTP synthase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.