missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CU CPT 22, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective toll-like receptor 1/2 (TLR1/2) inhibitor (IC50 = 0.58 μM). Exhibits no activity at TLR2/TLR6, TLR3, TLR4 and TLR7 or a panel of 10 representative kinases. Inhibits the release of proinflammatory cytokines TNF-α and IL-1β.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 362.37 |
Percent Purity | >98% |
Chemical Name or Material | CU CPT 22 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction