Learn More
Sartorius Cubis™ II Glass Draft Shield
Glass Draft Shield accessory for Cubis™ II
294.00€ - 367.00€
Specifications
For Use With (Equipment) | Cubis™ II High-Capacity Micro Balance |
---|---|
Material | Glass |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
17917330
|
Sartorius
YCCDSF |
Front Panel |
294.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17927340
|
Sartorius
YCCDSL |
Left Door |
294.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17907330
|
Sartorius
YCCDSR |
Right Door |
294.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17967330
|
Sartorius
YCCDSU |
Cover Slide |
367.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
Description
- Glass Draft Shield for use with Cubis™ II High-Capacity Micro Balance
Specifications
Cubis™ II High-Capacity Micro Balance | |
Glass |