Learn More
Sartorius Cubis II MTS Balance
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis™ II balance to all weighing tasks.
7086.00€ - 13980.00€
Specifications
Connection Type | D-sub connector, 9-pin |
---|---|
Display Type | Color Touchscreen |
Model | Cubis II MTS Balance |
Type | Balance |
Electrical Requirements | 00 to 240 VAC (±10%) |
Product Code | Brand | Capacity | Description | Readability | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Description | Readability | Price | Quantity & Availability | |||
30052872
|
Sartorius
MCE8202S-0-MTS |
8200 g | MCE Display with factory-installed essential weighing applications | 10 mg |
7086.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||
30052870
|
Sartorius
MCA8202S-0-MTS |
8200 g | MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions | 10 mg |
10250.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||
30052885
|
Sartorius
MCA125S-U-MTS |
120 g | MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions | 0.01 |
13980.00€
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||
Description
- Leading performance.
- Monolithical weighing system, integrated climate sensors, individual sample holders
- Error-free operation.
- Individual QApp workflows, motorized auto-leveling
- The modular weighing system Cubis™ II can be individually configured from different components.
Specifications
D-sub connector, 9-pin | |
Cubis II MTS Balance | |
00 to 240 VAC (±10%) |
Color Touchscreen | |
Balance |