Learn More
Thermo Scientific™ Culti-Loops™ Kirby Bauer Susceptibility QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4653004
545.84 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Culti-Loops™ Kirby Bauer Susceptibility Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures
.Includes:
Escherichia coli ATCC™ 25922™†
Escherichia coli ATCC™ 35218™†
Pseudomonas aeruginosa ATCC™ 27853™†
Staphylococcus aureus supsp. aureus ATCC™ 25923™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.

Specifications
25922TM* | |
Culti-Loops | |
Organism Sets | |
5 Loops/Organism |
Kirby Bauer Susceptibility QC Set/4 | |
Culti-Loop | |
Qualitative QC |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
102719 | Certificate of Analysis | 16/04/2024 | R4653004 |
821334 | Certificate of Analysis | 16/04/2024 | R4653004 |
702977 | Certificate of Analysis | 16/04/2024 | R4653004 |
561705 | Certificate of Analysis | 16/04/2024 | R4653004 |
543048 | Certificate of Analysis | 16/04/2024 | R4653004 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.