Learn More
WTW™ Cuvette Cells for PhotoLab™ Photometers
WTW™ Cells for PhotoLab™ Photometers are ideal for routine water and other analytics in the UV-VIS and VIS range.
Brand: WTW™ 250606
162.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Cells with matt side surface are suitable for the automatic cell detection of the photoLab™ filter and spectral photometer
- All glass and quartz cuvettes have placement marks to locate them in the photometer in the same orientation for even higher precision
- High quality optical glass for VIS measurements, high quality quartz glass for UV-VIS
Specifications
Cuvette | |
Reusable | |
10 mm | |
Rectangle |
REK 10 | |
Quartz | |
Photometers |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
WTW™ Cuvette Cells for PhotoLab™ Photometers > Path Length: 10 mm