missing translation for 'onlineSavingsMsg'
Learn More

CYM 5541, Tocris Bioscience™

Product Code. 30001687
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective S1P3 receptor allosteric agonist

Selective sphingosine-1-phosphate receptor 3 (S1P3) allosteric agonist (EC50 = 72-132 nM). Occupies a different space within the ligand binding pocket of S1P3 than S1P. Displays no activity at S1P1, S1P2, S1P4 and S1P5 receptors at concentrations up to 10 μM. Shows no significant activities in a profiling panel of 55 GPCRs, ion channels and transporters.

Chemical Identifiers

CAS 945128-26-7
Molecular Formula C19H28N2O2
Molecular Weight (g/mol) 316.445
InChI Key NDKGACIWVAOUQH-UHFFFAOYSA-N
Synonym n,n-dicyclohexyl-5-cyclopropyl-3-isoxazolecarboxamide, n,n-dicyclohexyl 5-cyclopropylisoxazol-3-yl carboxamide
PubChem CID 17253208
IUPAC Name N,N-dicyclohexyl-5-cyclopropyl-1,2-oxazole-3-carboxamide
SMILES C1CCC(CC1)N(C2CCCCC2)C(=O)C3=NOC(=C3)C4CC4

Specifications

Quantity 10 mg
Formula Weight 316.44
Percent Purity >97%
Chemical Name or Material CYM 5541
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'