Learn More
Cytiva Cytodex 3 Gamma Microcarriers
The microcarrier of choice for cells that are difficult to culture in vitro - particularly for cells with an epithelial-like morphology.
1339.00€ - 41700.00€
Specifications
For Use With (Application) | Production of large quantities of cells, viruses and recombinant cell products, studies on cell adhesion, differentiation and cell function |
---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15747207
|
Cytiva
17548801 |
30 g |
1339.00€
30g |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
15767207
|
Cytiva
17548803 |
3 kg |
41700.00€
3kg |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
15757207
|
Cytiva
17548802 |
300 g |
5974.00€
300g |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
Description
- Cytodex 3 Gamma is designed by chemically coupling of a thin layer of denatured porcine collagen type 1 to the cross-linked dextran matrix.
- As the collagen surface layer can be digested by a variety of proteolytic enzymes, harvest of such cells from the microcarriers can be performed with maintained high cell viability and membrane integrity.
- This feature of Cytodex 3 Gamma is beneficial in the development of successful serial subcultivation protocols for scaling up culture volumes.
- Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, differentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving cell transfer and studies on uptage of labelled compounds
Specifications
Production of large quantities of cells, viruses and recombinant cell products, studies on cell adhesion, differentiation and cell function |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-9.9 per cent of the mixture consists of component(s) of unknown acute toxicity.