missing translation for 'onlineSavingsMsg'
Learn More
D-Cycloserine, 98+%, Thermo Scientific Chemicals Product Code.: 11409136

D-Cycloserine, 98+%, Thermo Scientific Chemicals

Product Code. 11409136
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11409136

Brand: Thermo Scientific Alfa Aesar A18000.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-cycloserine, CAS # 68-41-7, is an amino acid analog of the amino acid D-alanine with broad-spectrum antibiotic activity.

Cycloserine is an amino acid analog antibiotic used for the treatment of tuberculosis and rarely used for the treatment of noninfectious diseases. It acts as a competitive inhibitor of D-alanine during peptidoglycan synthesis. It inhibits bacterial cell wall biosynthesis and prevents the conversion of D-Alanine to L-Alanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D-cycloserine is an analog of the amino acid D-alanine produced by Streptomyces garyphalus or S. orchidaceus and shows antibacterial activity
  • The activity of this compound is related to the ability to interfere with bacterial cell wall synthesis by competitively inhibiting two enzymes, L-alanine racemase (responsible for the formation of D-alanine from L-alanine) and D-alanine ligase (the enzyme responsible for the incorporation of D-alanine into the pentapeptide required for peptidoglycan formation and bacterial cell wall synthesis). This interaction impairs the peptidoglycan formation, which is needed for bacterial cell wall synthesis

Application

  • Depending on the concentration and the organism’s susceptibility, this compound may present bactericidal or bacteriostatic activities
  • Despite its antibiotic activity, this compound also has an excitatory activity
  • It has been described that D-cycloserine can bind to the glycine binding site of the NMDA receptor in the central nervous system
  • This compound presents an inhibitory activity against GABA transaminase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-41-7
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00005353
InChI Key DYDCUQKUCUHJBH-UWTATZPHSA-N
Synonym d-cycloserine, cycloserine, seromycin, orientomycin, oxamycin, cyclorin, cyclo-d-serine, cicloserina, farmiserina, miroseryn
PubChem CID 6234
ChEBI CHEBI:40009
IUPAC Name (4R)-4-amino-1,2-oxazolidin-3-one
SMILES C1C(C(=O)NO1)N

Specifications

Melting Point 137°C (decomposition)
Beilstein 80798
Merck Index 14,2751
Solubility Information Soluble in water; Slightly soluble in methanol and propylene glycol; Insoluble in chloroform and ether.
Quantity 5 g
Formula Weight 102.09
Optical Rotation +115° (c=2 in Water)
Percent Purity ≥98%
Chemical Name or Material D-Cycloserine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM29L009 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA18000.03,A18000.06
Lot Number10253246 Certificate TypeCertificate of Analysis Date09/12/2024 Catalog NumberA18000.03,A18000.06
Lot NumberU22K016 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberA18000.03,A18000.06
Lot Number10250676 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA18000.03,A18000.06
Lot NumberP13K028 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog NumberA18000.03,A18000.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Cycloserine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.