missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Histidine, 99%

Product Code. 11400117
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is known to inhibit cell division, and is also used by certain types of bacteria (such as Escherichia coli) as a source of L-Histidine. It can be used in the design of peptide drugs, cationic peptides, such as analogues of carnosine and also be used as a heavy metal sequestration agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is known to inhibit cell division, and is also used by certain types of bacteria (such as Escherichia coli) as a source of L-Histidine. It can be used in the design of peptide drugs, cationic peptides, such as analogues of carnosine and also be used as a heavy metal sequestration agent.

Solubility
Soluble in water and 1 M HCl (50 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351-50-8
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00065963
InChI Key HNDVDQJCIGZPNO-RXMQYKEDSA-N
Synonym d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid
PubChem CID 71083
ChEBI CHEBI:27947
IUPAC Name (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES N[C@H](CC1=CN=CN1)C(O)=O

Specifications

Melting Point ∼285°C (decomposition)
Beilstein 84089
Solubility Information Soluble in water and 1 M HCl (50mg/ml).
Quantity 25 g
Formula Weight 155.16
Optical Rotation +39° (c=5 in Water)
Percent Purity 99%
Chemical Name or Material D-Histidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.