missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dantrolene sodium hemiheptahydrate, Thermo Scientific™
86.20€ - 261.00€
Specifications
Chemical Name or Material | Dantrolene sodium hemiheptahydrate |
---|---|
CAS | 24868-20-0 |
Synonym | 1-[(5-(p-Nitrophenyl)-furfurylidene)amino]hydantoin sodium salt |
Solubility | slightly soluble aq. soln. |
Melting Point | 225°C to 226°C |
Description
Specifications
Dantrolene sodium hemiheptahydrate | |
1-[(5-(p-Nitrophenyl)-furfurylidene)amino]hydantoin sodium salt | |
225°C to 226°C | |
Life science | |
Crystalline Powder | |
≥97.5% |
24868-20-0 | |
slightly soluble aq. soln. | |
No special requirements | |
C14 H9 N4 Na O5 · 3.5 H2O | |
Yellow to Orange to Brown |
Safety and Handling
Recommended Storage : Freezer -20°C