missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DCUN1D1 293T Cell Transient Overexpression Lysate (Denatured)

Denatured cell transient overexpression lysate of DCUN1D1 293T.

Brand:  Abnova™ H00054165-T01.100uL

Product Code. 16100483

  • 215.00€ / 100µL
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Transient overexpression cell lysate was tested with Anti-DCUN1D1 antibody (H00054165-B01) by Western Blots.
Specifications

Specifications

293T
Gene ID: 54165
pCMV-DCUN1D1 full-length
100 μL
Human
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NP_065691.2
Transient overexpression cell lysate was tested with Anti-DCUN1D1 antibody by Western Blots
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.