Learn More
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
Brand:  Trajan Scientific and Medical™ 062448
 View more versions of this product
Product Code. 12965751
Quantity Price
1 94.95€ / 5m
10 71.40€ / 5m
Estimated Shipment: 28-01-2025
Description
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
  • Chemically inert and thermally stable up to 380°C
  • Suitable for organic and aqueous solvents
  • Ideal for biotechnology applications
Specifications
Specifications
Organic and Aqueous Solvents
1/Pk.
680 μm
Thermally stable up to 380°C
Tubing
530 μm
5 m