Learn More
Decon Laboratories™ Decomatic™ Cleaning Agent
Phosphate-free surface active cleaning agent. Decon Laboratories Decomatic™ Cleaning Agent can be use with laboratory glassware washing machines or for medical and specialized industrial applications.
Brand: Decon Laboratories™ DEC5
Beschreibung
Phosphate-free surface active cleaning agent. Decon Laboratories Decomatic™ Cleaning Agent can be use with laboratory glassware washing machines or for medical and specialized industrial applications.
- Free from phosphates, enzymes, EDTA or chlorine
- Biodegradable
- Bactericidal
- Non-flammable
- Non-foaming
- Totally rinsable
- Suitable for laboratory glassware, ceramics, plastic (except polycarbonate), rubber, Stainless Steel and iron metals
- Amphoteric surfactants and stabilizers
Spezifikation
13 | |
100.8°C | |
Polyethylene containers |
Pale Amber | |
A blend of amphoteric surface active agents, non-phosphate detergent builders, alkalis and sequester |
Sicherheit und Handhabung
- DECON DECOMATIC
Signalwort
- Warning
Gefahrenkategorie
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Gefahrenhinweis
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Ergänzende Informationen
- MIXTURE LIST-Contains : sodium hydroxide
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.