missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ deconex 16 PLUS

Liquid cleaning concentrate for the manual cleaning of laboratory glassware and utensils in immersion and ultrasonic baths. deconex 16 PLUS shows an excellent material compatibility especially with alkali- and corrosion-sensitive objects.

Brand:  Borer Chemie™ 509700.00-K05W

Product Code. 17406502

  • 137.00€ / 5kg
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Concentrate for manual cleaning
  • pH-neutral, phosphate-free, without EDTA, NTA and chlorine
  • Especially for alkali-sensitive materials
  • High yield, typical concentration: 30-50 mL/L
  • Rinses off without leaving residues
  • For consistent and flawless results
  • Wide cleaning spectrum
  • Technical support by the manufacturer
Specifications

Specifications

Liquid
Surfactants, complexing agents, sequestering agents
Canister
1 x 5 kg
4.7 L
5 kg
∼7.9 (1% Solution)
Concentrate
5°C to 25°C
1.2 gal.
11 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.