Learn More
Borer Chemie™ deconex 25 ORGANACID
Cleaner and neutralizer for laboratory washers. Shortens rinsing phases and reduces water consumption. deconex 25 ORGANACID is phosphate-free and suitable for environmental and water laboratories. Pre-rinsing agent against acid-soluble residues.
Brand:  Borer Chemie™ 502100.00-K25R
Additional Details : CAS : 77-92-9 Weight : 25.00000kg
 View more versions of this product
Product Code. 10014490
  • 296.80€ / 25kg
Estimated Shipment: 08-08-2024
Description
  • Neutralizer and cleaner for automated use
  • Ecological solution without phosphates and surfactants
  • Neutralizes carried-over alkaline residues, reduces rinsing phases
  • Reduces the consumption of rinsing water
  • Typical application concentration: 3 mL/L
  • Excellent material compatibility, also for aluminium
  • Can be rinsed off without leaving residues
  • Also as pre-rinsing agent against acid-soluble deposits
  • Technical support by the manufacturer
Specifications
Specifications
Liquid
Organic acid
Canister
1 x 25 kg
21.3 L
25 kg
Approx. 2.6 (1% Solution)
Concentrate
5°C to 25°C
5.6 gal.
55.1 lb.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • deconex 25 ORGANACID
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.