Learn More
Delta-Like Protein 4 Rat anti-Mouse, BUV563, Clone: 9A1.5, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 748395
531.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Delta-Like Protein 4 Monoclonal antibody specifically detects Delta-Like Protein 4 in Mouse samples. It is validated for Flow Cytometry
Specifications
Delta-Like Protein 4 | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
Delta like-4; Delta4; Dll4; DL4; DL-4 | |
Mouse DL4 Extracellular Domain Recombinant Protein | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
9A1.5 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.