Learn More
Smeg™ Demineralized Water Pump
Booster pump for non-pressurized, demineralized water.
Brand:  Smeg™ PAD
Product Code. 12366023
  • 271.00€ / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Booster pump for non-pressurized, demineralized water.
  • Allows feeding machine with demineralized water drawn from a nonpressurized tank positioned at a minimum height of 85 cm from ground
Specifications
Specifications
Booster Pump
Nonpressurized Water Tank