Learn More
R&D Systems™ Detergent Reagent
R&D Systems™ Detergent Reagent is supplied ready to use and is for use with cell proliferation assays.
Brand: R&D Systems™ 4890-25-02
Additional Details : Weight : 0.29100kg
Description
- Do not use past expiration date.
- Supplied ready for use.
- If the Detergent Reagent has been stored at 4°C, warm the bottle for 5 minutes at 37°C then invert gently while mixing to avoid frothing.
- Contains SDS which is an irritant. In case of contact, immediately flush eyes or skin with copious amounts of water. If swallowed, wash out mouth with water provided person is conscious.
Specifications
Store the unopened product at room temperature | |
250 mL |