Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
With sealed vacuum space between the inner and outer shells to provide superior performance and insulation provides maximum holding times.
Brand:  Taylor Wharton™ 4LDB
 View more versions of this product
Product Code. 11758519
  • 795.00€ / Each
Estimated Shipment: 09-09-2025
Specifications
Cryogenic Storage Dewar
1.1 gal.
4 L
10 Days
Certificates
View accessory products
Product Title
Taylor Wharton™ LD Series Cryogenic Storage Dewar > Capacity: 4 L