Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
With sealed vacuum space between the inner and outer shells to provide superior performance and insulation provides maximum holding times.
Brand:  Taylor Wharton™ 35LDB
 View more versions of this product
Product Code. 11788519
  • 1220.00€ / Each
Estimated Shipment: 09-09-2025
Specifications
Cryogenic Storage Dewar
9.2 gal.
35 L
152 Days
Documents
One moment while we fetch your results.
Certificates
View accessory products
Product Title
Taylor Wharton™ LD Series Cryogenic Storage Dewar > Capacity: 35 L