Learn More
Scotch-Brite™ Diamond Floor Pads, Purple
Diamond-coated floor pads for cleaning and polishing marble, terrazzo, slate, and polished concrete flooring. Helps eliminate floor finish and reduce labor costs.
Brand: Scotch-Brite™ 7100159510
This item is not returnable.
View return policy
Description
- Simultaneously cleans and polishes flooring to achieve an outstanding shine
- Ideal for marble, terrazzo, vinyl, slate, and polished concrete floors
- Easy to use: no expensive or harmful chemicals needed, only water
- Recommended for use with low and standard speed machines
Specifications
Cleaning Pads | |
305 mm | |
Marble, Terrazzo, Slate, and Polished Concrete Flooring | |
Round |
Purple | |
Burnishing, Cleaning | |
Diamond-coated |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Scotch-Brite™ Diamond Floor Pads, Purple > 305 mm