missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyryl-cAMP, sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Cell-permeable analog of cAMP; activates cAMP-dependent protein kinases. Promotes differentiation of dopaminergic neurons (iDA) from hPSCs in combination with other reagents.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 491.37 |
Percent Purity | >98% |
Chemical Name or Material | Dibutyryl-cAMP, sodium salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction