Learn More
Usbeck Laborgeraete™ Die-cast Zinc Burette Clamps
Brand: Usbeck Laborgeraete™ 2841
This item is not returnable.
View return policy
Specifications
Each | |
Die Cast Zinc, Nickel Plated, Plastic Coated | |
1 Burette |
Burette Clamp | |
roller grips | |
0 to 40 mm |
Product Content Correction
Product Title
Usbeck Laborgeraete™ Die-cast Zinc Burette Clamps > Clamping range: 0 to 40mm; For 1 Burette