missing translation for 'onlineSavingsMsg'
Learn More
Diethyl acetamidomalonate, 98+%, Thermo Scientific Chemicals Product Code.: 11427956

Diethyl acetamidomalonate, 98+%, Thermo Scientific Chemicals

Product Code. 11427956
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11427956

Brand: Thermo Scientific Alfa Aesar A14048.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-90-2
Molecular Formula C9H15NO5
Molecular Weight (g/mol) 217.221
MDL Number MFCD00009146
InChI Key ISOLMABRZPQKOV-UHFFFAOYSA-N
Synonym diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl esterShow MoreShow Less
PubChem CID 14041
IUPAC Name diethyl 2-acetamidopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)NC(=O)C

Specifications

Melting Point 96°C to 99°C
Boiling Point 185°C (20 mmHg)
Quantity 100 g
Beilstein 783883
Solubility Information Soluble in chloroform and methanol. Slightly soluble in water.
Formula Weight 217.22
Percent Purity ≥98%
Chemical Name or Material Diethyl acetamidomalonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243177 Certificate TypeCertificate of Analysis Date05/09/2022 Catalog NumberA14048.22,A14048.14,A14048.36
Lot Number10232307 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14048.22,A14048.14,A14048.36
Lot Number10174317 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14048.22,A14048.14,A14048.36
Lot Number10215332 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14048.22,A14048.14,A14048.36
Lot Number10178508 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14048.22,A14048.14,A14048.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl acetamidomalonate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.