missing translation for 'onlineSavingsMsg'
Learn More
Diethyl ethoxymethylenemalonate, 98+%, Thermo Scientific Chemicals Product Code.: 11464834

Diethyl ethoxymethylenemalonate, 98+%, Thermo Scientific Chemicals

Product Code. 11464834
1000 g
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
1000 g
This item is not returnable. View return policy

Product Code. 11464834

Brand: Thermo Scientific Alfa Aesar A13776.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl ethoxymethylenemalonate is used for synthesis of pyrido[3,2-e]pyrimido[1,2-c]pyrimidines. Initially, it was used to monitor lysine decarboxylase activity. It is useful in the determination of amino acids by precolumn derivatization by using reversed-phase high-performance liquid chromatography (HPLC). It plays an important role in the Gould-Jacobs reaction to prepare quinolines. For example, anile reacts with this reagent to give 4-hydroxyquinoline. It acts as an intermediate in the production of flumequine, which is a fluoroquinolone antibiotic.

Solubility
Miscible with chloroform, dichloromethane and methanol. Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-13-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00009148
InChI Key LTMHNWPUDSTBKD-UHFFFAOYSA-N
Synonym diethyl ethoxymethylenemalonate, diethyl 2-ethoxymethylene malonate, ethoxymethylenemalonic acid diethyl ester, diethyl ethoxymethylene malonate, diethyl emme, propanedioic acid, ethoxymethylene-, diethyl ester, diethylethoxymethylenemalonate, 1,3-diethyl 2-ethoxymethylidene propanedioate, diethyl ethoxymethylidene propanedioate
PubChem CID 6871
IUPAC Name diethyl 2-(ethoxymethylidene)propanedioate
SMILES CCOC=C(C(=O)OCC)C(=O)OCC

Specifications

Melting Point -33°C
Density 1.08
Boiling Point 136°C to 139°C (10 mmHg)
Flash Point 144°C (291°F)
Odor Ester-like
Refractive Index 1.462
Quantity 1000 g
Solubility Information Miscible with chloroform,dichloromethane and methanol. Immiscible with water.
Formula Weight 216.23
Percent Purity ≥98%
Chemical Name or Material Diethyl ethoxymethylenemalonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10233301 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13776.18,A13776.22,A13776.30,A13776.36,A13776.0B
Lot NumberGBFA007563 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13776.18,A13776.22,A13776.30,A13776.36,A13776.0B
Lot NumberI3953A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13776.18,A13776.22,A13776.30,A13776.36,A13776.0B
Lot NumberE22Z025 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13776.18,A13776.22,A13776.30,A13776.36,A13776.0B
Lot Number10212789 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13776.18,A13776.22,A13776.30,A13776.36,A13776.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl ethoxymethylenemalonate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.