Learn More
Faithful™ Differential Blood Cell Counter
Manual incremental counter with a durable ABS plastic outer shell. Faithful™ Differential Blood Cell Counter is ideal for classifying and counting white blood cells.
Brand: Faithful™ DBC6
This item is not returnable.
View return policy
Description
- ABS outer shell can resist collision.
- Color charts of white blood cells are supplied.
- Bell sound at 100% total record count.
- Reset knob clears figures.
- Quality certification: ISO 9001, ISO 13485
Specifications
5 Keys | |
250 x 80 x 50mm | |
Totalizer |