man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Materials Testing Instruments
Dimensional Analyzers
Mitutoyo™ Digital Caliper
Print
Mitutoyo™ Digital Caliper
POA - 2017.50€
See promotional offers below
Specifications
Display
LCD
View More Specs
Products
2
Documents
Product Certifications
Products
2
Product Code
Brand
Range
Price
Quantity & Availability
Product Code
Brand
Range
Price
Quantity & Availability
11776414
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Mitutoyo™
50050110
0 to 600 mm
2017.50€
Each
View alternative products
Save Up to
»
Estimated Shipment: 09-12-2024
Log in
to see stock.
Add to basket
11882633
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Mitutoyo™
500458
0 to 200 mm
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
View alternative products
Save Up to
»
Specifications
Display
LCD
Documents
Product Certifications
Back to Top