Learn More
Afora™ Dimroth Condenser
Provide additional surface area for highly efficient cooling with the Afora™ Dimroth Compressor, which pressurizes and heats refrigerant.
Brand:  Afora™ 1602/E
 View more versions of this product
Product Code. 12080088
  • 249.00€ / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cold finger
  • Coiled
  • Fluid-cooled
  • Internal double spiral
  • Often used with rotary evaporators
Specifications
Specifications
UNE 400316
29/32
0.01 kg
Dimroth Condenser