Learn More
Diphenylamine, ACS reagent, Thermo Scientific Chemicals Product Code.: 10269890
Diphenylamine, ACS reagent, Thermo Scientific Chemicals
Product Code. 10269890
500 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Product Code. 10269890
Brand: Thermo Scientific Acros 423655000
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00003014
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scaldShow MoreShow Less
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2
Specifications
Melting Point 52.0°C to 54.0°C
Color White
Boiling Point 302.0°C
Flash Point 152°C
Ignition Residue 0.03% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C6H5)2NH
Quantity 500 g
Beilstein 12, 174
Sensitivity (To nitrate) Passes Test
Merck Index 15, 3347
Solubility Information (in alcohol) Passes test
Formula Weight 169.23
Grade ACS Reagent
Physical Form Crystalline Flakes
Chemical Name or Material Diphenylamine, For analysis ACS
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464949 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number423651000,423655000,423650050
Lot NumberA0457333 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number423651000,423655000,423650050
Lot NumberA0448861 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog Number423651000,423655000,423650050
Lot NumberA0393453 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number423651000,423655000,423650050
Lot NumberA0438047 Certificate TypeCertificate of Analysis Date26/01/2022 Catalog Number423651000,423655000,423650050
Product Title
Diphenylamine, ACS reagent, Thermo Scientific Chemicals >