missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Direct Thermal Printable Labels, 25 mm Core

Printable paper labels for thermal transfer printers.

Brand:  Brady™ 236743

 View more versions of this product

Product Code. 11513135

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Direct thermal paper
  • Ideal for warehouse marking
Specifications

Specifications

Not Autoclavable
White
Direct Thermal Paper
Non-sterile
BBP™11 Label Printer, BBP™12 Industrial Label Printer, THT 76 mm Core Printers
18,000 Labels
0.099 mm
30.5 mm
Permanent Acrylic
Matte
Direct Thermal
Warehouse Marking
1
Rectangle
24.6 mm
21.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.