Learn More
Fisherbrand™ Dispenser tips
Robust, stable, and durable dispenser tips for several brands of pipettors. For dispensing of fluids with high viscosity and high vapor pressure.
Brand:  Fisherbrand™ 702620
48.70 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 15183646
  • 64.10€ / Pack of 50
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
In Stock
Add to basket
Description
Description
  • These disposable tips are suitable for the following multiple dispensers:
  • BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch/ S
  • Eppendorf Multipette™ 4780
  • Gilson Distriman™ and Gilson Repetman™
  • Ritter Ripette™ pro
  • EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
  • STARLAB RepeatOne™
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
ISO 8655
Dispenser Tip
Polypropylene
Non-sterile
For dispensing of fluids with high viscosity, high vapor pressure
Single-use
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24165
    Expires2025-06-30
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/06/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Product Title
Fisherbrand™ Dispenser tips > Capacity: 25mL