Learn More
Thermo Scientific Chemicals Disposable IR Cards, 5x10cm, KBr window, 19mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
137.00€ - 386.93€
Specifications
Description Disposable IR Cards
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15426575
View Documents
Thermo Scientific Chemicals
043935.KJ
5 Pk. This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
15436575
View Documents
Thermo Scientific Chemicals
043935.KM
25 Pk.
386.93€
Pack of 25
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Size: 5 x 10cm
  • KBr window
  • 19mm aperture
Specifications
Specifications
Disposable IR Cards
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Title
Thermo Scientific Chemicals Disposable IR Cards, 5x10cm, KBr window, 19mm aperture