Learn More
3M™ Disposable Particulate Respirator, 8000 Series
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
29.37€ - 62.25€
Specifications
Design Type | Cup/Molded |
---|---|
Disposable | Single-use |
Product Type | General Purpose Particulate Respirator |
Sterility | Non-sterile |
FDA Cleared Surgical Mask | No |
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Quantity | Price | Quantity & Availability | |||||
12302399
|
3M™
7000006980 |
FFP1 NR D | Yes | 10/Pk. |
34.80€
Pack of 10 |
Estimated Shipment: 20-06-2025
Log in to see stock. |
|||||
12382389
|
3M™
7000034734 |
FFP1 NR D | No | 20/Pk. |
34.80€
Pack of 20 |
In Stock
Log in to see stock. ![]() |
|||||
11713363
|
3M™
7100003010 |
FFP2 NR D | No | 1 Ea. |
60.65€
Pack of 20 |
In Stock
Log in to see stock. ![]() |
|||||
11723363
|
3M™
7000006980 |
FFP1 NR D | Yes | 10/Pk. | N/A | ||||||
11736003
|
3M™
7100010817 |
FFP2 NR D | Yes | 10/Pk. |
62.25€
Pack of 10 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
11733363
|
3M™
7100101091 |
FFP2 | Yes | 10/Pk. |
39.50€
Pack of 10 |
In Stock
Log in to see stock. ![]() |
|||||
11703363
|
3M™
7000034734 |
FFP1 NR D | No | 20/Pk. | N/A | ||||||
Description
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.Specifications
Cup/Molded | |
General Purpose Particulate Respirator | |
No |
Single-use | |
Non-sterile |
Product Title
3M™ Disposable Particulate Respirator, 8000 Series