Learn More
Rogo Sampaic™ Dissection Needle
Brand:  Rogo Sampaic™ FMD001
 View more versions of this product
Product Code. 11711436
  • 63.00€ / Pack of 10
Estimated Shipment: 12-05-2025
Specifications
Dissection needle
Straight
40 mm
Stainless Steel
125 mm
Documents
One moment while we fetch your results.
Product Title
Rogo Sampaic™ Dissection Needle > Length: 125mm