Learn More
Rogo Sampaic™ Dissection Needle
Brand:  Rogo Sampaic™ FMD003
 View more versions of this product
Product Code. 11778541
  • 59.00€ / Pack of 10
Estimated Shipment: 28-04-2025
Specifications
Dissection needle
Stainless Steel
Lanceolate
146 mm
Product Title
Rogo Sampaic™ Dissection Needle > Length: 146mm