Learn More
Kimble Chase™ Distilling Flasks
Brand:  Kimble Chase™ 2947500010
 View more versions of this product
Product Code. 11875850
  • 171.00€ / Each
Estimated Shipment: 24-11-2025
Specifications
10 mL
Borosilicate Glass
Pear
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kimble Chase™ Distilling Flasks > Capacity: 10mL