Learn More
Kimble Chase™ Distilling Flasks
Brand:  Kimble Chase™ 2955100025
 View more versions of this product
Product Code. 11895850
  • 196.00€ / Each
Estimated Shipment: 08-08-2025
Specifications
25 mL
Round
Round
SDS
Documents Certificates
Product Title
Kimble Chase™ Distilling Flasks > Capacity: 25mL